2,6-Dichloro-3-formylphenylboronic acid
Supplier: Apollo Scientific
|
Warning
|
OR14152-1G
OR14152-5G
APOSOR14152-1GEA
29
EUR
APOSOR14152-1G
APOSOR14152-5G
2,6-Dichloro-3-formylphenylboronic acid
2,6-Dichloro-3-formylphenylboronic acid
Formula:
C₇H₅BCl₂O₃ Melting Pt: 150…154 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06796322 CAS Number: 1218790-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...