α-Oxothiophen-2-acetic acid 96%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Oxo-2-(thiophen-2-yl)acetic acid
OR14231-100G
OR14231-25G
OR14231-5G
APOSOR14231-100GEA
768
EUR
APOSOR14231-100G
APOSOR14231-25G
APOSOR14231-5G
α-Oxothiophen-2-acetic acid 96%
α-Oxothiophen-2-acetic acid
Formula:
C₆H₄O₃S MW: 156,1600 g/mol Melting Pt: 88…95 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005441 CAS Number: 4075-59-6 EINECS: 223-794-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...