3-Iodoanisole 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-iodoanisole
OR14264-100G
APOSOR14264-100GEA
84.1
EUR
APOSOR14264-100G
3-Iodoanisole 98%
3-Iodoanisole
Formula:
C₇H₇IO MW: 234,04 g/mol Boiling Pt: 243…244 °C Density: 1,78 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001048 CAS Number: 766-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...