7-Cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-7-cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidine

OR14278-1G OR14278-500MG
APOSOR14278-1GEA 288 EUR
APOSOR14278-1G APOSOR14278-500MG
7-Cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
7-Cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Formula: C₈H₉N₅
MW: 175,19 g/mol
Melting Pt: 235…237 °C
Storage Temperature: Ambient
MDL Number: MFCD07366548
CAS Number: 885949-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR