Thiazol-2-ylmethanol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Hydroxymethyl)thiazole, 2-Thiazolemethanol
OR14307-5G
OR14307-25G
APOSOR14307-5GEA
79.9
EUR
APOSOR14307-5G
APOSOR14307-25G
Thiazol-2-ylmethanol
Thiazol-2-ylmethanol
Formula:
C₄H₅NOS Melting Pt: 62…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200855 CAS Number: 14542-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...