Quinhydrone

Supplier: Apollo Scientific
Danger

Synonyms: Benzoquinone-hydroquinone, p-Benzoquinone hydroquinone complex, 2,5-Cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol (1:1)

OR14340-100G OR14340-250G OR14340-500G
APOSOR14340-100GEA 52.2 EUR
APOSOR14340-100G APOSOR14340-250G APOSOR14340-500G
Quinhydrone
Quinhydrone
Formula: C₆H₄(OH)₂·C₆H₄O₂
MW: 218,21 g/mol
Melting Pt: 173…174 °C
Density: 1,4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010310
CAS Number: 106-34-3
EINECS: 203-387-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR