Succinic acid ACS
OR14341-1KG
OR14341-25KG
OR14341-5KG
APOSOR14341-1KGEA
184
EUR
APOSOR14341-1KG
APOSOR14341-25KG
APOSOR14341-5KG
Succinic acid ACS
Succinic acid
Formula:
HOOCCH₂CH₂COOH MW: 118,09 g/mol Boiling Pt: 235 °C Melting Pt: 188…190 °C Density: 1,564 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002789 CAS Number: 110-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...