Methyl-6-amino-5-bromonicotinate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 6-amino-5-bromonicotinate
OR14385-1G
OR14385-5G
OR14385-500MG
APOSOR14385-1GEA
23.4
EUR
APOSOR14385-1G
APOSOR14385-5G
APOSOR14385-500MG
Methyl-6-amino-5-bromonicotinate
Methyl 6-amino-5-bromonicotinate
Formula:
C₇H₇BrN₂O₂ MW: 231,05 g/mol Melting Pt: 166…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659561 CAS Number: 180340-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...