N-(3-Methoxyphenyl)urea
Supplier: Apollo Scientific
|
Warning
|
OR14387-1G
OR14387-500MG
OR14387-5G
APOSOR14387-1GEA
176
EUR
APOSOR14387-1G
APOSOR14387-500MG
APOSOR14387-5G
N-(3-Methoxyphenyl)urea
N-(3-Methoxyphenyl)urea
Formula:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Melting Pt: 138…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025423 CAS Number: 139-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...