3,5-Dichloro-4-methoxybenzenecarboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 3,5-Dichloro-4-methoxybenzoic acid

OR14433-1G OR14433-250MG
APOSOR14433-1GEA 26.3 EUR
APOSOR14433-1G APOSOR14433-250MG
3,5-Dichloro-4-methoxybenzenecarboxylic acid
3,5-Dichloro-4-methoxybenzenecarboxylic acid
Formula: C₈H₆Cl₂O₃
MW: 221,04 g/mol
Melting Pt: 199…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00210404
CAS Number: 37908-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR