2-Methylquinoline-6-carboxylic acid 99%

Supplier: Apollo Scientific
Warning

OR14488-100G OR14488-25G OR14488-5G
APOSOR14488-100GEA 98.6 EUR
APOSOR14488-100G APOSOR14488-25G APOSOR14488-5G
2-Methylquinoline-6-carboxylic acid 99%
2-Methylquinoline-6-carboxylic acid
Formula: C₁₁H₉NO₂
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00748504
CAS Number: 635-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR