(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone 97%

Supplier: Apollo Scientific

OR14534-10G OR14534-25G
APOSOR14534-10GEA 75.4 EUR
APOSOR14534-10G APOSOR14534-25G
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone 97%
(R)-(-)-4-Benzyl-3-propionyl-2-oxazolidinone
Formula: C₁₃H₁₅NO₃
MW: 233,2631 g/mol
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00269687
CAS Number: 131685-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR