N-Boc-4-methyl-L-phenylalanine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Boc-p-Me-Phe-OH, Boc-Phe(4-Me)-OH
OR14572-1G
OR14572-5G
APOSOR14572-1GEA
94.5
EUR
APOSOR14572-1G
APOSOR14572-5G
N-Boc-4-methyl-L-phenylalanine
N-Boc-4-methyl-L-phenylalanine
Formula:
C₁₅H₂₁NO₄ MW: 279,336 g/mol Melting Pt: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD01317027 CAS Number: 80102-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...