Boc-4-methyl-D-phenylalanine 98%

Supplier: Apollo Scientific
Warning

Synonyms: Boc-D-Phe(4-Me)-OH, Boc-p-Me-D-Phe-OH

OR14573-1G OR14573-5G
APOSOR14573-1GEA 88.7 EUR
APOSOR14573-1G APOSOR14573-5G
Boc-4-methyl-D-phenylalanine 98%
Boc-4-methyl-D-phenylalanine
Formula: C₁₅H₂₁NO₄
MW: 279,336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317726
CAS Number: 80102-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR