Methyl-3-iodo-1H-indole-6-carboxylate 95%
Supplier: Apollo Scientific
|
Warning
|
OR1459-1G
OR1459-250MG
OR1459-500MG
OR1459-5G
APOSOR1459-1GEA
353
EUR
APOSOR1459-1G
APOSOR1459-250MG
APOSOR1459-500MG
APOSOR1459-5G
Methyl-3-iodo-1H-indole-6-carboxylate 95%
Methyl-3-iodo-1H-indole-6-carboxylate
Formula:
C₁₀H₈INO₂ MW: 301,09 g/mol Melting Pt: 150 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD06797504 CAS Number: 850374-98-0 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...