Fmoc-3,4-dichloro-D-phenylalanine 95+%

Supplier: Apollo Scientific

OR14609-5G OR14609-1G
APOSOR14609-5GEA 170 EUR
APOSOR14609-5G APOSOR14609-1G
Fmoc-3,4-dichloro-D-phenylalanine 95+%
Fmoc-3,4-dichloro-D-phenylalanine
Formula: C₂₄H₁₉Cl₂NO₄
Storage Temperature: Ambient
MDL Number: MFCD00273472
CAS Number: 177966-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR