N-Boc-3-(2-thienyl)-L-alanine
Supplier: Apollo Scientific
|
Warning
|
OR14642-1G
OR14642-5G
APOSOR14642-1GEA
27.7
EUR
APOSOR14642-1G
APOSOR14642-5G
N-Boc-3-(2-thienyl)-L-alanine
N-Boc-3-(2-thienyl)-L-alanine
Formula:
C₁₂H₁₇NO₄S Melting Pt: 71…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062051 CAS Number: 56675-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...