Fmoc-D-allylglycine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
D-2-Allylglycine, N-FMOC protected, Fmoc-D-Gly(allyl)-OH
OR14673-5G
APOSOR14673-5GEA
114
EUR
APOSOR14673-5G
Fmoc-D-allylglycine 98%
Fmoc-D-allylglycine
Formula:
C₂₀H₁₉NO₄ MW: 337,38 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01311776 CAS Number: 170642-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...