(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95+%

Supplier: Apollo Scientific

Synonyms: (1R,3S)-(+)-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected, (1R,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

OR14679-5G
APOSOR14679-5GEA 119 EUR
APOSOR14679-5G
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95+%
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229,276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320858
CAS Number: 161660-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR