2-Chloro-L-phenylalanine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
H-Phe(2-Cl)-OH
OR14687-25G
APOSOR14687-25GEA
185
EUR
APOSOR14687-25G
2-Chloro-L-phenylalanine
2-Chloro-L-phenylalanine
Formula:
C₉H₁₀ClNO₂ MW: 199,6 g/mol Melting Pt: 196…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077921 CAS Number: 103616-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...