4-Chloro-L-phenylalanine 97%

Supplier: Apollo Scientific
Danger

Synonyms: L-4-Chlorophenylalanine, H-Phe(4-Cl)-OH

OR14691-10G OR14691-5G OR14691-1G
APOSOR14691-10GEA 58 EUR
APOSOR14691-10G APOSOR14691-5G APOSOR14691-1G
4-Chloro-L-phenylalanine 97%
4-Chloro-L-phenylalanine
Formula: C₉H₁₀ClNO₂
MW: 199,6 g/mol
Melting Pt: 263 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00063065
CAS Number: 14173-39-8
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR