3,4-Dichloro-D-phenylalanine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(R)-2-Amino-3-(3,4-dichlorophenyl)propanoic acid, H-3,4-Dichloro-D-Phe-OH, H-D-Phe(3,4-DiCl)-OH, 2-Amino-3-3-4-dichlorophenyl-propanoic-acid, D-3,4-Dichlorophenylalanine
OR14720-5G
OR14720-10G
OR14720-25G
APOSOR14720-5GEA
143
EUR
APOSOR14720-5G
APOSOR14720-10G
APOSOR14720-25G
3,4-Dichloro-D-phenylalanine
3,4-Dichloro-D-phenylalanine
Formula:
C₉H₉Cl₂NO₂ MW: 234,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00671682 CAS Number: 52794-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...