3-(Aminomethyl)phenol
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Hydroxybenzylamine
OR14753-1G
OR14753-5G
OR14753-250MG
APOSOR14753-1GEA
62.5
EUR
APOSOR14753-1G
APOSOR14753-5G
APOSOR14753-250MG
3-(Aminomethyl)phenol
3-(Aminomethyl)phenol
Formula:
C₇H₉NO Melting Pt: 165…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00798977 CAS Number: 73604-31-6 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...