2-Hydroxy-4-methoxy-5-nitropyridine
Supplier: Apollo Scientific
|
Warning
|
OR14756-1G
OR14756-250MG
APOSOR14756-1GEA
128
EUR
APOSOR14756-1G
APOSOR14756-250MG
2-Hydroxy-4-methoxy-5-nitropyridine
2-Hydroxy-4-methoxy-5-nitropyridine
Formula:
C₆H₆N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD07367779 CAS Number: 607373-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...