2-(3-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde 95+%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-(1,3-Dioxoprop-2-yl)-2-nitrobenzoic acid
OR1483-1G
OR1483-25G
OR1483-5G
APOSOR1483-1GEA
136
EUR
APOSOR1483-1G
APOSOR1483-25G
APOSOR1483-5G
2-(3-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde 95+%
2-(3-Hydroxycarbonyl-2-nitrophenyl)malondialdehyde
Formula:
C₁₀H₇NO₆ MW: 237,17 g/mol Melting Pt: 215…217 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216502 CAS Number: 205680-83-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...