5-Amino-3-isopropyl-1,2,4-thiadiazole

Supplier: Apollo Scientific
Warning

OR14831-1G OR14831-250MG
APOSOR14831-1GEA 0 EUR
APOSOR14831-1G APOSOR14831-250MG
5-Amino-3-isopropyl-1,2,4-thiadiazole
5-Amino-3-isopropyl-1,2,4-thiadiazole
Formula: C₅H₉N₃S
Storage Temperature: Ambient
MDL Number: MFCD03093824
CAS Number: 32039-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR