2,4,5-Triamino-6-hydroxypyrimidine sulphate

Supplier: Apollo Scientific
Warning

Synonyms: 2,5,6-Triaminopyrimidin-4(1H)-one sulphate, 4-Hydroxy-2,5,6-triaminopyrimidine sulphate, 6-Hydroxy-2,4,5-triaminopyrimidine sulphate

OR14903-100G OR14903-1KG
APOSOR14903-100GEA 64 EUR
APOSOR14903-100G APOSOR14903-1KG
2,4,5-Triamino-6-hydroxypyrimidine sulphate
2,4,5-Triamino-6-hydroxypyrimidine sulphate
Formula: C₄H₉N₅O₅S
MW: 239,21 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012785
CAS Number: 35011-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR