2-Morpholino-1,3-thiazole-5-carbaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
OR14933-1G
OR14933-5G
OR14933-10G
APOSOR14933-1GEA
84.4
EUR
APOSOR14933-1G
APOSOR14933-5G
APOSOR14933-10G
2-Morpholino-1,3-thiazole-5-carbaldehyde 97%
2-Morpholino-1,3-thiazole-5-carbaldehyde
Formula:
C₈H₁₀N₂O₂S MW: 198,25 g/mol Melting Pt: 162,5…164.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01568825 CAS Number: 1011-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...