3,4,5-Trichloropyridazine 99%

Supplier: Apollo Scientific
Warning

OR14946-1G OR14946-25G OR14946-5G
APOSOR14946-1GEA 21.9 EUR
APOSOR14946-1G APOSOR14946-25G APOSOR14946-5G
3,4,5-Trichloropyridazine 99%
3,4,5-Trichloropyridazine
Formula: C₄HCl₃N₂
Boiling Pt: 117…118 °C/14mmHg
Melting Pt: 54…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00006464
CAS Number: 14161-11-6
EINECS: 238-006-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR