3-Chloro-6-(4-chlorophenyl)pyridazine

Supplier: Apollo Scientific
Warning

OR14960-1G OR14960-250MG
APOSOR14960-1GEA 96 EUR
APOSOR14960-1G APOSOR14960-250MG
3-Chloro-6-(4-chlorophenyl)pyridazine
3-Chloro-6-(4-chlorophenyl)pyridazine
Formula: C₁₀H₆Cl₂N₂
Melting Pt: 194…204 °C
Storage Temperature: Ambient
MDL Number: MFCD06801340
CAS Number: 58059-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR