1-Butanesulphonyl chloride 98%

Supplier: Apollo Scientific
Danger

Synonyms: 1-Butanesulphonylchloride

OR14981-100G
APOSOR14981-100GEA 82.8 EUR
APOSOR14981-100G
1-Butanesulphonyl chloride 98%
1-Butanesulphonyl chloride
Formula: C₄H₉ClO₂S
MW: 156,63 g/mol
Boiling Pt: 80…81 °C
Density: 1,2 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00007464
CAS Number: 2386-60-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR