1,3-Bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione

Supplier: Apollo Scientific
Warning

Synonyms: 1,3-Bis(hydroxymethyl)-5,5-dimethylhydantoin

OR14987-10G OR14987-5G
APOSOR14987-10GEA 37.9 EUR
APOSOR14987-10G APOSOR14987-5G
1,3-Bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione
1,3-Bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione
Formula: C₇H₁₂N₂O₄
MW: 188,18 g/mol
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00467199
CAS Number: 6440-58-0
EINECS: 229-222-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR