3-Bromo-2-chlorothiophene 98%
Supplier: Apollo Scientific
|
Warning
|
OR15031-100G
OR15031-25G
APOSOR15031-100GEA
275
EUR
APOSOR15031-100G
APOSOR15031-25G
3-Bromo-2-chlorothiophene 98%
3-Bromo-2-chlorothiophene
Formula:
C₄H₂BrClS MW: 197,48 g/mol Boiling Pt: 70 °C/9mmHg Density: 1,808 g/cm³ (25 °C) Flash Pt: 102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043883 CAS Number: 40032-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...