1-[(4-Nitrophenyl)sulphonyl]tetrahydro-4(1H)-pyridinone

Supplier: Apollo Scientific
Warning

Synonyms: 1-[(4-Nitrophenyl)sulphonyl]piperidin-4-one

OR15052-1G OR15052-5G OR15052-500MG
APOSOR15052-1GEA 234 EUR
APOSOR15052-1G APOSOR15052-5G APOSOR15052-500MG
1-[(4-Nitrophenyl)sulphonyl]tetrahydro-4(1H)-pyridinone
1-[(4-Nitrophenyl)sulphonyl]tetrahydro-4(1H)-pyridinone
Formula: C₁₁H₁₂N₂O₅S
MW: 284,29 g/mol
Melting Pt: 199…201 °C
Storage Temperature: Ambient
MDL Number: MFCD08689704
CAS Number: 924869-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR