N-(Chloroacetyl)morpholine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-(Chloroacetyl)morpholine, 4-(2-Chloroacetyl)morpholine
OR1511-100G
OR1511-25G
OR1511-5G
APOSOR1511-100GEA
382
EUR
APOSOR1511-100G
APOSOR1511-25G
APOSOR1511-5G
N-(Chloroacetyl)morpholine 98%
N-(Chloroacetyl)morpholine
Formula:
C₆H₁₀ClNO₂ MW: 163,61 g/mol Melting Pt: 26…30 °C Density: 1,257 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00721939 CAS Number: 1440-61-5 EINECS: 215-880-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...