3-Oxo-3-pyridin-3-ylpropionitrile 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Oxo-3-(3-pyridyl)propanenitrile
OR15117-1G
OR15117-5G
APOSOR15117-1GEA
84.1
EUR
APOSOR15117-1G
APOSOR15117-5G
3-Oxo-3-pyridin-3-ylpropionitrile 97%
3-Oxo-3-pyridin-3-ylpropionitrile
Formula:
C8H6N2O MW: 146,150 g/mol Melting Pt: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD08282797 CAS Number: 30510-18-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...