1-Chloro-3-iodo-2-nitrobenzene

Supplier: Apollo Scientific
Warning

OR15151-1G OR15151-25G OR15151-5G
APOSOR15151-1GEA 21.9 EUR
APOSOR15151-1G APOSOR15151-25G APOSOR15151-5G
1-Chloro-3-iodo-2-nitrobenzene
1-Chloro-3-iodo-2-nitrobenzene
Formula: C₆H₃ClINO₂
Melting Pt: 60…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08443947
CAS Number: 937601-42-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR