Methyl (4-bromophenyl)acetate 97%

Supplier: Apollo Scientific
Warning

Synonyms: methyl 2-(4-bromophenyl)acetate

OR15214-100G OR15214-25G OR15214-500G
APOSOR15214-100GEA 88.7 EUR
APOSOR15214-100G APOSOR15214-25G APOSOR15214-500G
Methyl (4-bromophenyl)acetate 97%
Methyl (4-bromophenyl)acetate
Formula: C₉H₉BrO₂
MW: 229,0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00126844
CAS Number: 41841-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR