1-Methyl-1H-indazol-5-amine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Amino-1-methyl-1H-indazole, 1-methyl-1H-indazole-5-amine, 1-METHYL-1H-INDAZOL-5-AMINE
OR15216-1G
OR15216-250MG
APOSOR15216-1GEA
24.8
EUR
APOSOR15216-1G
APOSOR15216-250MG
1-Methyl-1H-indazol-5-amine
1-Methyl-1H-indazol-5-amine
Formula:
C₈H₉N₃ Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD03305455 CAS Number: 50593-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...