2-[5-Methyl-2-(2-thienyl)-1,3-thiazol-4-yl]acetic acid 95%

Supplier: Apollo Scientific
Warning

OR15235-100MG OR15235-250MG
APOSOR15235-100MGEA 50.9 EUR
APOSOR15235-100MG APOSOR15235-250MG
2-[5-Methyl-2-(2-thienyl)-1,3-thiazol-4-yl]acetic acid 95%
2-[5-Methyl-2-(2-thienyl)-1,3-thiazol-4-yl]acetic acid
Formula: C₁₀H₉NO₂S₂
MW: 239,32 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08443972
CAS Number: 924868-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR