3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid

Supplier: Apollo Scientific
Warning

OR15242-1G OR15242-250MG
APOSOR15242-1GEA 234 EUR
APOSOR15242-1G APOSOR15242-250MG
3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid
3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid
Formula: C9H7NO3S
Melting Pt: 287…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00456558
CAS Number: 272437-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR