6-Bromo-1H-indole-2-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 6-Bromoindole-2-carboxylic acid

OR15252-1G OR15252-25G OR15252-5G
APOSOR15252-1GEA 53.9 EUR
APOSOR15252-1G APOSOR15252-25G APOSOR15252-5G
6-Bromo-1H-indole-2-carboxylic acid
6-Bromo-1H-indole-2-carboxylic acid
Formula: C₉H₆BrNO₂
MW: 240,05 g/mol
Melting Pt: 221…223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02664469
CAS Number: 16732-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR