2-Amino-5-bromo-3-(hydroxymethyl)pyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-5-bromo-3-pyridinemethanol
OR15266-10G
OR15266-25G
OR15266-5G
APOSOR15266-10GEA
299
EUR
APOSOR15266-10G
APOSOR15266-25G
APOSOR15266-5G
2-Amino-5-bromo-3-(hydroxymethyl)pyridine
2-Amino-5-bromo-3-(hydroxymethyl)pyridine
Formula:
C₆H₇BrN₂O Melting Pt: 137…140 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05664025 CAS Number: 335031-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...