4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(Thiomorpholin-4-yl)benzaldehyde
OR15298-1G
OR15298-5G
APOSOR15298-1GEA
219
EUR
APOSOR15298-1G
APOSOR15298-5G
4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde
4-(1,4-Thiazinan-4-yl)benzenecarbaldehyde
Formula:
C₁₁H₁₃NOS MW: 207,3 g/mol Melting Pt: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276967 CAS Number: 27913-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...