2'-Formylbiphenyl-4-carboxylic acid ethyl ester
Supplier: Apollo Scientific
Synonyms:
Ethyl 2'-formyl-[1,1'-biphenyl]-4-carboxylate
OR15335-1G
OR15335-500MG
OR15335-5G
APOSOR15335-1GEA
270
EUR
APOSOR15335-1G
APOSOR15335-500MG
APOSOR15335-5G
2'-Formylbiphenyl-4-carboxylic acid ethyl ester
2'-Formylbiphenyl-4-carboxylic acid ethyl ester
Formula:
C₁₆H₁₄O₃ Melting Pt: 45…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276944 CAS Number: 885950-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...