1H-Indol-3-ylhydrosulphide

Supplier: Apollo Scientific
Warning

Synonyms: 3-Sulphanyl-1H-indole, 1H-Indole-3-thiol

OR15338-1G OR15338-250MG
APOSOR15338-1GEA 184 EUR
APOSOR15338-1G APOSOR15338-250MG
1H-Indol-3-ylhydrosulphide
1H-Indol-3-ylhydrosulphide
Formula: C₈H₇NS
MW: 149.22 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00152110
CAS Number: 480-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR