4-methoxy-1H-indazol-3-amine 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Amino-4-methoxy-1H-indazole, 4-Methoxy-1H-indazol-3-amine
OR15340-1G
OR15340-5G
OR15340-25G
APOSOR15340-1GEA
32.1
EUR
APOSOR15340-1G
APOSOR15340-5G
APOSOR15340-25G
4-methoxy-1H-indazol-3-amine 95%
4-methoxy-1H-indazol-3-amine
Formula:
C₈H₉N₃O MW: 163,18 g/mol Melting Pt: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276946 CAS Number: 886362-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...