Methyl-3-iodoimidazo[1,2-a]pyridine-6-carboxylate 95%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 3-iodoimidazo[1,2-a]pyridine-6-carboxylate

OR15353-500MG OR15353-1G
APOSOR15353-500MGEA 62.5 EUR
APOSOR15353-500MG APOSOR15353-1G
Methyl-3-iodoimidazo[1,2-a]pyridine-6-carboxylate 95%
Methyl 3-iodoimidazo[1,2-a]pyridine-6-carboxylate
Formula: C₉H₇IN₂O₂
MW: 302,0686 g/mol
Melting Pt: 188…191 °C
Storage Temperature: Ambient
MDL Number: MFCD07021385
CAS Number: 460087-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR