2-Chloro-N-(tetrahydro-2-furanylmethyl)acetamide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N-(Chloroacetyl)-2-(aminomethyl)tetrahydrofuran
OR15399-1G
OR15399-5G
OR15399-10G
APOSOR15399-1GEA
174
EUR
APOSOR15399-1G
APOSOR15399-5G
APOSOR15399-10G
2-Chloro-N-(tetrahydro-2-furanylmethyl)acetamide
2-Chloro-N-(tetrahydro-2-furanylmethyl)acetamide
Formula:
C₇H₁₂ClNO₂ Melting Pt: 42…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD01182294 CAS Number: 39089-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...