2-Hydroxy-4'-(methylsulphonyl)acetophenone

Supplier: Apollo Scientific
Warning

OR15411-1G OR15411-250MG
APOSOR15411-1GEA 316 EUR
APOSOR15411-1G APOSOR15411-250MG
2-Hydroxy-4'-(methylsulphonyl)acetophenone
2-Hydroxy-4'-(methylsulphonyl)acetophenone
Formula: C₉H₁₀O₄S
Melting Pt: 136…138 °C
Storage Temperature: Ambient
CAS Number: 197240-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR